missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK beta 2/PRKAB2 Antibody (OTI4H4), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71226PCP
This item is not returnable.
View return policy
Description
AMPK beta 2/PRKAB2 Monoclonal antibody specifically detects AMPK beta 2/PRKAB2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| AMPK beta 2/PRKAB2 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI4H4 | |
| Western Blot | |
| 5'-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5'-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, protein kinase, AMP-activated, beta 2 non-catalytic subunit | |
| Full length human recombinant protein of human PRKAB2 (NP_005390) produced in E.coli. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5565 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction