missing translation for 'onlineSavingsMsg'
Learn More

Amphotericin B, Antibiotic Solution, MP Biomedicals

Product Code. p-24634 Shop All MP Biomedicals Products
Click to view available options
Quantity:
20 mL
50 mL
Unit Size:
20mL
50mL
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Product Code. 11410435

Brand: MP Biomedicals 091672346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Antibiotic solution Concentration: 250μg/mL Effective against yeast and fungi

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1397-89-3
Molecular Formula C47H73NO17
Molecular Weight (g/mol) 924.09
MDL Number MFCD00877763
InChI Key APKFDSVGJQXUKY-ZNVUZQDLSA-N
Synonym Fungizone
PubChem CID 134129663
IUPAC Name (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
SMILES C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O

Specifications

Type Amphotericin B
Quantity 20 mL
Chemical Name or Material Amphotericin B
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.