missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Ultra Pure, 1kg, MP Biomedicals

Product Code. 11491632 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11491632

Brand: MP Biomedicals 04808211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • Fe: <<0.5 ppm
  • Mg: <<0.13 ppm
  • Pb: <<0.1 ppm
  • Mn: <<0.06 ppm
  • Ca: <<0.1 ppm
  • Zn: <<0.5 ppm
  • Cu: <<0.21 ppm
  • O.D. (260nm) of a saturated solution: <<0.1

TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-20-2
Quantity 1 kg
Molecular Formula H8N2O4S
MDL Number MFCD00003391
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=O
IUPAC Name sulfuric acid diamine
Molecular Weight (g/mol) 132.13
PubChem CID 6097028
ChEBI CHEBI:62946
Grade Ultrapure
Chemical Name or Material Ammonium Sulfate, Ultra Pure
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.