missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Standard Solution, Ion HIQU, Chem-Lab
Click to view available options
Quantity:
100 mL
500 mL
Description
Ammonium standard solution is used as an analytical/elemental standard and in ion chromatography. It is used in ammonia determination and to calibrate equipment with ammonia sensors.
Specifications
Specifications
Concentration | 2.97g NH4Cl per 1L of H2O |
Quantity | 500 mL |
Color | Colorless |
SMILES | [NH4+].[Cl-] |
Formula Weight | 53.49g/mol |
Odor | Odorless |
Density | 1.00g/mL |
pH | ±5 |
Solubility | Soluble |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'