missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Standard Solution, Ion HIQU, Chem-Lab

Artikelnummer. 11969012
Click to view available options
Quantity:
100 mL
500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Contains 1000μg/mL of NH4+. Ammonia Standard Solution, Ion HIQU, Chem-Lab is a single component solution with 2.97g NH4Cl per 1L of H2O.

Ammonium standard solution is used as an analytical/elemental standard and in ion chromatography. It is used in ammonia determination and to calibrate equipment with ammonia sensors.

Chemische Identifikatoren

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride

Spezifikation

Concentration 2.97g NH4Cl per 1L of H2O
Quantity 100 mL
Color Colorless
SMILES N.Cl
Formula Weight 53.49g/mol
Odor Odorless
Density 1.00g/mL
pH ±5
Solubility Soluble
Physical Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'