missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride-Ammonia Buffer, pH 10.1, Chem-Lab
Click to view available options
Quantity:
1000 mL
Description
This buffer solution helps to maintain an alkaline pH level. This buffer is made from a weak base (ammonia) and one of its salts (ammonium sulfate).
Specifications
Specifications
Melting Point | 0°C |
Boiling Point | 100°C |
Color | Colorless |
Density | 1.01g/mL |
pH | 10.1 |
Physical Form | Liquid |
Quantity | 1000 mL |
Solution Type | Buffer |
Formulation | 6.75g NH4Cl + 57mL NH4OH 25% per 1L H2O |
Odor | Odorless |
Show More |
Safety and Handling
Product Identifier
- Ammonium chloride-Ammonia buffer pH 10.1
- Danger
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : NH4OH, NH4Cl
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'