Learn More
WTW™ Ammonia Gas-sensitive Combination Electrode
The half cells of the 500 series for which you will need a reference electrode type R 503 (an exception for this is the gas-sensitive NH 500/2 with integrated reference electrode).
Brand:  WTW™ 106395
Product Code. 10461402
  • 1215.00€ / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
3 Replacement Membrane Heads