missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution, 1M, Chem-Lab

Product Code. 11975517
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for laboratory use and titrations. Ammonia Solution, 1M, Chem-Lab is a solution of ammonia in water, with 35.05g NH4OH per 1L H2O. It can be used to study chromatography, base solutions, analytical reagents, titration and volumetric solutions.

Ammonium hydroxide solution can be used to study chromatography, base solutions, analytical reagents, titration and volumetric solutions.

  • Aqueous ammonia is used as a complexant and base.
  • It can dissolve silver oxide residues.
  • When mixed with dilute hydrogen peroxide in the presence of a metal ion, the peroxide decomposes rapidly.
  • Aqueous ammonia can be used as a precursor to some alkyl amines.
  • Ammonia solution is found in household and jewelry cleaning agents, and has many industrial and commercial applications.

Specifications

CAS 1336-21-6
Molecular Formula H5NO
Concentration 1M
MDL Number MFCD00066650
Density 0.98g/mL
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 1 L
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.05g/mol
Concentration or Composition (by Analyte or Components) 35.05g NH4OH per 1L H2O
Normality 1 N (±0.002, 20°C)
Solvent or Matrix Water
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Ammonia 1 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : ammonia
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'