missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Amidine Latex Beads, 4% w/v, 0.2 μm

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene.

Brand:  Molecular Probes™ A37314

380.00 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 11524067

  • 410.00€ / 15mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

This positively-charged hydrophobic amidine latex is particularly suited for the preparation of latex intermediates. The only surface functional group present on the particle is amidine, which is not sensitive to the aggregating effects of polyvalent cations. These particles are sensitive to negatively charged contaminants or multivalent anions. The amidine latexes should be used in low to neutral pH environments. The amidine group is a base and with a pK ∼10-11.

Microspheres Are Available in Many Sizes and Surface Functionalities
• 25 sizes to choose from—0.02 microns to 16 microns
• 20 different surface—for ready conjugation to virtually any material
• Surfactant free—ultraclean
• Various quantities—from milliliter to 500-liter amounts

Key Applications of Microspheres
• Instrument calibration (flow cytometry, microscopy, HTS, HCS)
• Flow testing (microfluidics, blood flow, water flow, and air flow)
• Cell biology tracers (cell differentiation and cell tracing)
• Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

Get More Information
Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

We’ll Make a Custom Microsphere Product for You
If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C).
Latex Beads
15 mL
Amidine
Polystyrene
IDC
Room Temperature
0.2 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.