Learn More
Invitrogen™ Ambion™ 0.1 to 2kb RNA Ladder
Suitable for sizing single-stranded RNA from 0.1 to 2kb in glyoxal or formaldehyde agarose gels
Brand: Invitrogen™ 15623100
Includes: 0.1 to 2Kb RNA Ladder is supplied at 1μg/μL in 10mM HEPES (pH 7.2), 2mM EDTA
Description
0.1-2kb RNA Ladder consists of ten bands in even increments (0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.8, 1.0, 1.5, and 2.0kb) to simplify size estimation. The 0.3Kb band is more intense than the other bands, allowing easy orientation and identification of RNA bands on the gel. All RNA molecules in the ladder are polyA-tailed (40 nt) for convenient use as control templates for cDNA synthesis. Compatible with formaldehyde-containing, glyoxal-containing and polyacrylamide gels.
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Dry Ice
For Research Use Only. Not for use in diagnostic procedures.