Learn More
Emma Safety Footwear Amazone Safety Shoes
On top of being an extremely safe shoe, Amazone also offers its wearer exceptional comfort. The steel toe cap will protect from falling objects, while the easy twist sole provides a 'sticky grip'.
Brand:  Emma Safety Footwear MM33254649
 View more versions of this product
Product Code. 15978371
  • 139.00€ / 1 pair
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Black
Hydro-Tec™ Sanitized Silver
49
Laces
Amazone
Oiled Nubuck
Steel