Learn More
Fisherbrand™ Aluminum Weighing Boats, Pack of 100, Oil Free
GREENER_CHOICE
Dishes for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.
28.00€ - 90.00€
Specifications
Autoclavable Not Autoclavable
Bottom Shape Round
Disposable Single-use
Material Aluminum
Shape Circular
View More Specs
Products 9
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
30182871
GREENER_CHOICE
View Documents
Fisherbrand™
120932FRF
5 mL
39.00€
Pack of 100
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
30182875
GREENER_CHOICE
View Documents
Fisherbrand™
120930FRF
70 mL
90.00€
Pack of 100
In Stock
to see stock.
Edge
Add to basket
30182878
GREENER_CHOICE
View Documents
Fisherbrand™
120931FRF
25 mL
65.00€
Pack of 100
In Stock
to see stock.
Edge
Add to basket
30182868
GREENER_CHOICE
View Documents
Fisherbrand™
120935FRF
42 mL
28.00€
Pack of 100
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
30182874
GREENER_CHOICE
View Documents
Fisherbrand™
120936FRF
80 mL
60.00€
Pack of 100
In Stock
to see stock.
Edge
Add to basket
30182872
GREENER_CHOICE
View Documents
Fisherbrand™
120937FRF
140 mL
70.00€
Pack of 100
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
30182870
GREENER_CHOICE
View Documents
Fisherbrand™
HEA14521AFRF
20 mL
35.00€
Pack of 100
In Stock
to see stock.
Edge
Add to basket
30182877
GREENER_CHOICE
View Documents
Fisherbrand™
HEA14521BFRF
34 mL
36.00€
Pack of 100
In Stock
to see stock.
Edge
Add to basket
30182876
GREENER_CHOICE
View Documents
Fisherbrand™
HEA14521CFRF
50 mL
45.00€
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
The Aluminum Weighing Boat may be used for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.
  • These sturdy yet lightweight boats feature a smooth, flat bottom for sample handling.
  • Oil free
Passed through a rigorous, three-step cleansing process to remove forming oil or residue for more reliable weighing and moisture sample analysis.
  • Processed and packaged in clean room environment
  • Lot traceable
  • Volume: 70 mL
  • Dimensions: 2.8 x 0.7 in. (71 x 17 mm)
  • Pack of 100
  • Crinkle finish
  • Thickness: 0.075 mm
  • Certificates for Oil-Free Aluminum Boats available upon request
Specifications
Specifications
Not Autoclavable
Single-use
Circular
Weighing Boat
Round
Aluminum
Non-sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Aluminum Weighing Boats, Pack of 100, Oil Free