missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Standard Solution, WTW™

Product Code. 11704825
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
For few parameters such as nitrite, which are subject to a quick conversion, freshly prepared control standards of stock solutions versus the multi-parameter standard checks are mandatory. Stock solutions are also more cost-effective than the already confectioned CombiChecks™. With stock solutions, the correct dilution must be self-calculated and conducted. The use of standards is supported via AQA functions in the photoLab™ photometers.
  • Free chlorine
  • Bound chlorine
  • Formaldehyde
  • Hydrazine
  • Hydrogen peroxide
  • Hydrogen sulfide
  • Phenol
  • Silicon
  • Sulfide
  • Sulfite
  • Anionic surfactants
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1000mg/L
Quantity 500 mL
Color Colorless
Traceability to NIST Yes
Solution Contains Aluminum
pH 0.5
Boiling Point 100°C
Solubility Soluble in water
Physical Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'