missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Clamp for use with MV050 Vacuum Filtration Manifolds
Replacement aluminum clamp for use with MV050 Vacuum Filtration Manifolds
Brand: Cytiva 10440004
This item is not returnable.
View return policy
Description
- Product Type: Clamp
- Material: Aluminum
- For Use with (Equipment): MV050 (and GV050) Vacuum Filtration Manifolds
- For Use with (Applications): Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries)
Specifications
Autoclavable | |
MV050 & GV050 Manifold | |
Non-sterile | |
MV050/0/06 | |
MV050 |
Reusable | |
Aluminum | |
Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries) | |
1 Ea. | |
Clamp |