missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum with Over laminated Polyester Transport Signs

Product Code. 11720294
Click to view available options
Displayed Text:
Corrosive Substance 8
Environmentally Hazardous Substances
Miscellaneous hazardous substances 9
Organic Peroxide 5.2
Oxidizing substances and organic peroxides 5
Toxic and Harmful Substances 6
Warning of Toxic Substances
Thickness:
0.024 mm
0.27 mm
0.271 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11720294

Brand: Brady™ 241646

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number PIC 302-TRI 100-B7525
Material Aluminum, Polyester
Quantity 1 Ea.
Description Aluminum with Polyester Overlaminate
For Use With (Application) Pipe markers and safety signs
Thickness 0.27 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Warning of Toxic Substances
Product Type Transport Sign
Color Black on Yellow
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.