man
Test
chevron-down
Select Language
English
French
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Film and Foil Wrapping and Accessories
Film and Foil Wrapping
Korff™ Aluminum Foil
11962563
Print
Korff™ Aluminum Foil
Dimensions (L x W): 100mm x 100mm
Brand:
Korff™ 60275
View more versions of this product
Product Code.
11962563
73.75€
/ Pack of 1000
Quantity
Estimated Shipment: 07-11-2024
Log in
to see stock.
Explore more special offers
Specifications
Width (Metric)
100 mm
Length (Metric)
100 mm
Material
Aluminum