Learn More
VACUUBRAND™ Aluminum Clamping Rings
A rapid connection that can be mounted without any tools for soft seals and indium seals.
Brand: VACUUBRAND™ 20660001
18.54 EUR valid until 2025-04-18
Use promo code "24095" to get your promotional price.
Description
- Provided with wing nut
- Available in two different sizes
Specifications
20/25 mm | |
Aluminum | |
DN 20/25 mm |
For Small KF Flange | |
1 Each | |
Clamping Ring |
Promotions
-
Multiple Manufacturers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Product Content Correction
Product Title
VACUUBRAND™ Aluminum Clamping Rings > Size: DN 20/25mm