missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ALPL/CEN1p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1515.00€ - 2495.00€
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | alkaline phosphatase, liver/bone/kidney |
Gene Alias | AP-TNAP,FLJ40094,FLJ93059,HOPS,MGC161443,MGC167935,TNAP,TNSALP |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013355
|
Abnova™
FG0217.100uL |
100 μL |
1515.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16023355
|
Abnova™
FG0217.200uL |
200 μL |
2495.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
AP-TNAP,FLJ40094,FLJ93059,HOPS,MGC161443,MGC167935,TNAP,TNSALP | |
249 | |
Liquid |
Tube | |
alkaline phosphatase, liver/bone/kidney | |
ALPL | |
DNA/RNA |