missing translation for 'onlineSavingsMsg'
Learn More

alpha-(p-Toluenesulfonyl)-4-fluorobenzylisonitrile, 97%, Thermo Scientific™

Product Code. 15459915
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165806-95-1
Molecular Formula C15H12FNO2S
Molecular Weight (g/mol) 289.32
MDL Number MFCD03787932
InChI Key UXCQPEDHCCJBNL-UHFFFAOYNA-N
Synonym a-tosyl-4-fluorobenzyl isocyanide, 1-fluoro-4-isocyano tosyl methyl benzene, alpha-p-toluenesulfonyl-4-fluorobenzylisonitrile, a-p-toluenesulfonyl-4-fluorobenzylisonitrile, 4-fluorophenyl isocyano methyl 4-methylphenyl sulphone, alpha-4-methylphenyl sulphonyl-4-fluorobenzylisonitrile, alpha-tosyl-4-fluorobenzyl isocyanide, alpha-p-toluenesulfonyl-4-fluorobezylisonitrile, 1-4-fluorophenyl-1-tosyl methyl isocyanide
PubChem CID 10827289
IUPAC Name 1-[(4-fluorophenyl)-isocyanomethyl]sulfonyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C([N+]#[C-])C1=CC=C(F)C=C1

Specifications

Quantity 1 g
Formula Weight 289.32
Percent Purity 97%
Chemical Name or Material alpha-(p-Toluenesulfonyl)-4-fluorobenzylisonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.