missing translation for 'onlineSavingsMsg'
Learn More

alpha-Isopropylphenylacetic acid, 97%, Thermo Scientific Chemicals

Product Code. 11453087
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3508-94-9
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00021708
InChI Key HDLQGISFYDYWFJ-UHFFFAOYSA-N
Synonym alpha-isopropylphenylacetic acid, 2-isopropyl-2-phenylacetic acid, 3-methyl-2-phenyl-butyric acid, 3-methyl-2-phenylbutyric acid, butyric acid, 3-methyl-2-phenyl, benzeneacetic acid, alpha-1-methylethyl, 2-phenylisovaleric acid, acmc-1bn3m, benzeneacetic acid, alpha-1-methylethyl-9ci, alpha-isopropylphenylaceticacid
PubChem CID 19024
IUPAC Name 3-methyl-2-phenylbutanoic acid
SMILES CC(C)C(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 57°C to 62°C
Quantity 1 g
Beilstein 2045915
Solubility Information Slightly soluble in water.
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material alpha-Isopropylphenylacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'