missing translation for 'onlineSavingsMsg'
Learn More

Alloxan monohydrate, 98%

Product Code. 11488216
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
This item is not returnable. View return policy

Product Code. 11488216

Brand: Thermo Scientific Alfa Aesar A15324.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3237-50-1 | C4H4N2O5 | 160.085 g/mol

Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytotoxic compound that causes oxidative base damage to nuclear DNAAlloxan monohydrate is used as a precursor to prepare purple dye murexide. It is also used in research models to induce diabetes and destroy beta cells. Further, it is a strong oxidizing agent and it forms a hemiacetal with its reduced reaction product dialuric acid.

Solubility
Soluble in water, ethanol, acetone, glacial acetic acid and methanol. Slightly soluble in chloroform, petroleum ether, toluene, ethyl acetate and acetic anhydride. Insoluble in ether.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3237-50-1
Molecular Formula C4H4N2O5
Molecular Weight (g/mol) 160.085
MDL Number MFCD00149399
InChI Key ZIIHZVKHFWOENY-UHFFFAOYSA-N
Synonym barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6
PubChem CID 312231
IUPAC Name 5,5-dihydroxy-1,3-diazinane-2,4,6-trione
SMILES C1(=O)C(C(=O)NC(=O)N1)(O)O

Especificaciones

Melting Point ∼260°C (decomposition)
Beilstein 5309394
Merck Index 14,282
Formula Weight 160.09 (142.07 Anhydrous)
Percent Purity 98%
Quantity 50 g
Chemical Name or Material Alloxan monohydrate

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado