missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alda 1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Activator of aldehyde dehydrogenase 2 (ALDH2). Increases activity of wild-type ALDH2*1 and variant ALDH2*2 (by ∽2-fold and 11-fold respectively). Capable of partly restoring mutant ALDH2*2 activity; protective against cardiac ischemia.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 324.16 |
Percent Purity | >99% |
Chemical Name or Material | Alda 1 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'