Learn More
Thermo Scientific Chemicals Albumin, for biochemistry, from egg, powder
Albumin, for biochemistry, from chicken egg, powder.
28.94€ - 551.75€
Specifications
Name Albumin
pH Range 6 to 8
Source Egg
Form Powder
View More Specs
Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10636051
View Documents Promotion Details
Thermo Scientific Chemicals
400451000
100 g
48.54€
100g
10307131
View Documents Promotion Details
Thermo Scientific Chemicals
400450500
50 g
28.94€
50g
10720672
View Documents Promotion Details
Thermo Scientific Chemicals
400455000
500 g
150.75€
500g
11976941
View Documents Promotion Details
Thermo Scientific Chemicals
400450025
2.5 kg
551.75€
2.50kg
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Albumin, bovine serum (BSA) is available as a protease-free, lyophilized powder that may be stably stored at 2-8°c
• It is comprised of 583 amino acid residues, and has a molecular weight of 66.5 kDa
Applications
• BSA is used as a protein standard in quantification assays, such as the Bradford protein assay
• It is used to stabilize enzymes in restriction digestions in molecular biology experiments
• It is a blocking agent in immunoblots thus increasing signal to noise ratios by quenching non-specific immunochemical reactions
• BSA is a nutritive agent and used as a supplement in cell and microbial culture media
Specifications
Specifications
Albumin
6 to 8
Egg
Powder
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Albumin, egg, powder