Learn More
Emma Safety Footwear Alaska XD Safety Shoes
Offers exceptional comfort while the steel toe cap protects from falling objects. Easy twist sole provides a 'sticky grip'.
Brand:  Emma Safety Footwear MM30356645
 View more versions of this product
Product Code. 15898787
  • 133.00€ / 1 pair
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
Alaska XD
Oiled Nubuck
45