Learn More
Brady™ Aggressive Adhesive Continuous Multipurpose Nylon Label Tape for M6/M7 Printers
Aggressive adhesive and high pliability are the hallmarks of this nylon cloth label material designed to meet a broad array of identification requirements, from control panels to liquid nitrogen environments.
Brand:  Brady™ 174151
 View more versions of this product
Product Code. 17879054
  • 31.05€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Highly pliable material conforms tightly to curved surfaces such as wires and cables while also adhering to stainless steel, textured ABS, polypropylene and many other material types
  • Resists exposure to cold temperatures, chemicals, greases, oils, cleaners, and detergents
  • Low profile label material adheres to smooth, rough, or powder coated surfaces or even materials with low surface energy
  • UL Recognized to UL969, CSA to C222 No 015-95, RoHS compliant to 2011/65/EU (Pass 2015/863)
  • Made of B-499 nylon cloth material
Specifications
Specifications
Autoclavable
M6C-375-499
None
Matte
Nylon
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
30 Feet
-40°F to +194°F
0.0065 in.
30 ft. x 0.375 in.
9.14 m
9.53 mm
Halogen-free (DIN VDE 0472 Part 815)
Permanent Acrylic
White
Continuous
Single-side Printable
Labels
Component and Equipment Labeling, Data and Telecommunications Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Wire and Cable Labeling
No
Rectangle
-40°C to +90°C
0.166 mm
30 ft.
0.375 in.