missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose Low EEO

Molecular Biology Grade.

Brand:  Melford A20080-50.0

 View more versions of this product

Product Code. 30017977

  • 137.00€ / 50g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Molecular Biology Grade.

Molecular Biology Grade. High purity for preparative and analytical electrophoresis of nucleic acids >1000 bp.

Gelling Temperature:

34.5 -37.5°C

Sulfate:

0.15

EEO:

0.09- 0.13

DNA Binding:

None Detected

DNA and RNase:

None Detected

Specifications

Specifications

Agarose, Low EEO
9012-36-6
50 g
Molecular Biology
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions