missing translation for 'onlineSavingsMsg'
Learn More

AG 825, Tocris Bioscience™

Product Code. 18023942
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective ErbB2 inhibitor

Selective ErbB2 inhibitor (IC50 values are 0.15 and 19 μM at ErbB2 and ErbB1 respectively). Preferentially triggers p38 MAP kinase-dependent apoptosis in androgen-independent prostate cancer cells.

Chemical Identifiers

CAS 149092-50-2
Molecular Formula C19H15N3O3S2
Molecular Weight (g/mol) 397.47
MDL Number MFCD01074971
InChI Key KXDONFLNGBQLTN-UHFFFAOYSA-N
Synonym tyrphostin ag 825, tyrphostin c15, 4-hydroxy-3-methoxy-5-benzothiazolylthiomethyl benzylidenecyanoacetamide, 5-benzothiazol-2-yl thiomethyl-4-hydroxy-3-methoxybenzylidenecyanoacetamide, 2e-3-3-1,3-benzothiazol-2-ylsulfanyl methyl-4-hydroxy-5-methoxyphenyl-2-cyanoprop-2-enamide, tocris-1555, biomolki_000008, biomolki2_000018, bmk1-b8, tyrphostin ag
PubChem CID 6091659
ChEBI CHEBI:75405
IUPAC Name 3-{3-[(1,3-benzothiazol-2-ylsulfanyl)methyl]-4-hydroxy-5-methoxyphenyl}-2-cyanoprop-2-enamide
SMILES COC1=CC(C=C(C#N)C(N)=O)=CC(CSC2=NC3=CC=CC=C3S2)=C1O

Specifications

Quantity 10 mg
Formula Weight 397.47
Percent Purity >98%
Chemical Name or Material AG 825
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.