missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ AF330 EBC Model Color Scale Kit

Brand:  Lovibond™ 433300

Additional Details : Weight : 2.34000kg

Product Code. 11904159

  • 2275.00€ / Each
Estimated Shipment: 24-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Récupel-bijdrage uitgesloten

Description

Description

Specifications

Specifications

Color Scale Kit
Applications: Beers, Malts and Caramels, Food and Beverages, Household Products, Medical and Clinical, Petroleum Oils and Waxes, Edible Oils and Fats, Sugar Solutions, Syrups and Honey, Pharmaceuticals and Cosmetics, Textiles, Coatings, Plastics, Chemicals, Swimming Pools and Spas, Water and Waste Waters
AF330 EBC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions