Learn More
Mecmesin™ Advanced Force Gauge
The Advanced Force Gauge (AFG) is the most versatile, fully-featured digital instrument.
Brand:  Mecmesin™ 850421
Encompass
 View more versions of this product
Product Code. 11937979
  • 1950.00€ / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
With ever-increasing demands for improved quality, durability and ease-of-use of all products, there is a growing need for more physical testing during the manufacturing processes. The Advanced Force Gauge (AFG) is a powerful instrument packed full of innovative features designed to suit a breadth of tension and compression tests.
The AFG offers enhanced test features and data analysis capabilities:
  • Max and 1st Peak mode - identify critical load points from the test
  • Fast data-acquisition speed - captures break-force readings accurately
  • Audible and visual pass/fail alarms - alerts operators immediately when testing
  • Overload warning with trend bar - to minimize accidental damage to loadcell
  • In-built storage for up to 500 readings - test rapidly now and download results later
  • Data transfer via RS232, digimatic or analog - output data to PC or printer for further analysis
  • Connection capability to external load cells and torque sensors - becomes a versatile display for many different applications
Specifications
Specifications
2500 N
201 mm
70 mm
650 g