Learn More
Mecmesin™ Advanced Force Gauge
The Advanced Force Gauge (AFG) is the most versatile, fully-featured digital instrument.
Brand: Mecmesin™ 850421
Description
With ever-increasing demands for improved quality, durability and ease-of-use of all products, there is a growing need for more physical testing during the manufacturing processes. The Advanced Force Gauge (AFG) is a powerful instrument packed full of innovative features designed to suit a breadth of tension and compression tests.
The AFG offers enhanced test features and data analysis capabilities:
- Max and 1st Peak mode - identify critical load points from the test
- Fast data-acquisition speed - captures break-force readings accurately
- Audible and visual pass/fail alarms - alerts operators immediately when testing
- Overload warning with trend bar - to minimize accidental damage to loadcell
- In-built storage for up to 500 readings - test rapidly now and download results later
- Data transfer via RS232, digimatic or analog - output data to PC or printer for further analysis
- Connection capability to external load cells and torque sensors - becomes a versatile display for many different applications
Specifications
2500 N | |
201 mm | |
70 mm | |
650 g |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.