Learn More
Grant Instruments™ Advanced Air Cleaner Recirculator, 2 Lamps
Product Code. 17775149
Click to view available options
:
UK plug
Product Code. 17775149
Brand: Grant Instruments™ UVRMi UK
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Powerful/efficient UV cleaner-recirculator. Two UV lamps providing air disinfection in hospitals, research laboratories and clinics. The UV air flow cleaner-recirculator consists of 2 germicidal UV lamps, a fan, unit equipped with dust filters and a control unit, confined in a flow through chamber.
The inner mirror surface of the recirculator chamber reflects ultraviolet rays thereby increasing the density of UV radiation and enhancing the disinfection effect.
  • Two UV-lamps
  • Full user protection from direct UV radiation;
  • Two operation modes — short time (under timer control) and continuous operation
  • Digital control unit allows tracking of overall UV lamp operating time
  • 1 min-24 hours/non stop timer
Recirculators fixation:
  • Convenient fixation on walls (standard)
  • Mounting on a movable tripod (optional)
TRUSTED_SUSTAINABILITY
Specifications
Filter Efficiency 18 mW/cm2/sec, 14 m3/hour
Includes 2 Lamps
Mounting UVR-S
Type UV-Air Flow Cleaner-Recirculator