Learn More
Thermo Scientific™ ABgene™ Adhesive PCR foil seal
Brand:  Thermo Scientific™ ABgene™ AB-0626-RMP340
Product Code. 15383129
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • Sample is accessed by piercing foil with a pipettor tip
  • No adhesive build-up on tip after piercing
  • End tabs for easy seal application and removal
  • Recommended for use in conjunction with a thermal cycler that has a screw or clip down lid
  • Temperature range: -80°C to 150°C