Learn More
3M™ Adflo™ Powered Air Purifying Respirator System with Speedglas™ 9100 MP Welding Helmet
The Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ 9100 MP Welding Helmet is designed to help protect the eyes and face from radiation, sparks, and spatter.
Brand:  3M™ 7100054792_CLEAN
Product Code. 17895656
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • The flip-up functionality built into the 3M™ Speedglas™ 9100 MP Welding Helmet with 3M™ Adflo™ Powered Air Purifying Respirator System makes it an adaptable protection solution.
  • This system allows for quick swapping between welding filter and clear view, helping the user inspect their work while they go.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbo: approx. 7-9 hours || ADF: up to 1800 hours
COMBINATION
High Efficiency (HE)
One Size
No
ADF: Lithium 3V || Turbo: Lithium-ion
Black
Auto-darkening Filter
9100-Air Welding Helmet, HE Filter, Lithium Ion Battery, Auto-Darkening Filter