Learn More
3M™ Adflo™ Lightweight Breathing Tube
3M™ Adflo™ Lightweight Breathing Tube without QRS (Quick Release Swivel) is designed for use with the Speedglas welding helmet series 9000 and 3M™ Protective Visors.
Brand:  3M™ 7000000189
Product Code. 15719637
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • This breathing tube can also be used for the Speedglas welding helmet series 9100 and Versaflo headtops M-series, by adding the helmet breathing tube adapter (part no 53 35 05)
  • You can extend the service life of the breathing tube by using a tube cover or a heavy-duty rubber breathing tube