Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Socket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100112
 View more versions of this product
Product Code. 10772522
  • 42.00€ / Pack of 10
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sockets
  • Acc. to DIN 12249 form E
  • DIN standard
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Specifications
NS 12/21
1.8 mm
15 mm
Duran Glass