Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01100214
This item is not returnable.
View return policy
Description
- Cones
- Acc. to DIN 12249 form B
- Standard, conical ground cones
- Made of DURAN™ tubing
Specifications
NS 14/23 | |
1.5 mm | |
13 mm | |
Duran Glass |