missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Activated Penetratin™

Successfully used to internalize covalently attached peptides, oligonucleotides and siRNA.

Brand:  MP Biomedicals™ 11PENA0500

Product Code. 11427130

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Applications: Activated PenetratinTM 1 Peptide is a novel tool for peptide and siRNA delivery. It has been successfully used to internalize covalently attached peptides, oligonucleotides, and siRNA. It is sold ready for coupling to peptides or oligonucleotides containing thiol (-SH) functions.
Product Overview: PenetratinTM 1 is a patented 16-amino acid peptide corresponding to the third helix of the homeodomain of antennapedeia protein. This peptide is able to translocate across biological membranes by an energy-independent mechanism (1-3) and has been used successfully to internalize covalently attached peptides (4), oligonucleotides (5) and siRNA (6). Activated PenetratinTM 1 is sold ready for coupling to peptides or oligos containing thiol (-SH) functions. Biotinylated Activated PenetratinTM 1 possesses a biotin group at the C-terminal end allowing the detection of the internalized molecule with an appropriate avidin or streptavidin reagent.
Product Description: The Active PenetratinTM 1 Peptide Transfection reagent features a patented 16 amino acid peptide corresponding to the third helix of the homeodomain of the Antennapedia protein (pAntp) from Drosophila melanogaster.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stable for two years when frozen (<-15 °C).
Transfection
500 μg
Lyophilized Powder
2588
Reagents
Transfection Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.