missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, DARCO™, Honeywell Fluka™

Product Code. 15622580
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

DARCO™, −100 mesh particle size, powder

  • DARCO is a registered trademark of Norit Americas, Inc.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Mesh Size −100mesh particle size
Quantity 250 g
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Grade DARCO™
Chemical Name or Material Activated Charcoal
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'