Learn More
Cytiva ActiSM liquid medium
A chemically defined ADCF medium optimized for high-yield protein production. Designed for use with ActiPro medium.
Brand:  Cytiva SH31040.01
 View more versions of this product
Product Code. 15825261
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • HyClone ActiSM cell culture medium has been formulated to provide high yields of recombinant proteins in cell culture processes when using Chinese hamster ovary (CHO) cell lines.
  • The medium is chemically defined, animal-derived component-free (ADCF), and optimized for high-yield protein production.
  • ActiSM medium does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batc
  • ActiSM medium is intended for use in combination with HyClone ActiPro medium and Cell Boost™ 7a and Cell Boost 7b supplements as part of the ActiPro system of media and supplements designed with adaptation and production in mind.
  • The ActiPro system is suitable for general biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 to 8 °C, away from light
Sterile
Poloxamer 188
Bottle
Liquid
Sterile-filtered
No Glutamine
500 mL