missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Actinomycin D

Product Code. p-7180308
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 11588666

Brand: Invitrogen™ 11805017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • Acts by interfering with mRNA synthesis
  • ctinomycin D is used as a selective antibiotic for resistant bacteria, generally at a concentration of about 1μg/mL
  • Product is provided as a powder
  • Made into a stock solution at 1mg/ml in DMSO

Cell Analysis, Cell Structure, Cloning, Nucleus, Nucleoli & Nuclear Envelope, Transformation

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at facilities that comply with cGMP manufacturing requirements and that are certified to ISO 13485 and registered with FDA as medical device manufacturers.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 μg/mL
Form Powder
Product Type Antibiotic
Quantity 5 mg
compliance-icons compliance-icons
Product Identifier
  • Actinomycin D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1A
Hazard Statement
  • H300-Fatal if swallowed.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Actinomycin D

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.