missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ACT 462206
Potent, dual orexin receptor antagonist
183.00€ - 629.00€
Specifications
Purity | >99% |
---|---|
Product Type | ACT 462206 |
Molecular Weight (g/mol) | 388.48 |
Inhibitors | ACT 462206 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18770528
|
Tocris Bioscience™
5319/10 |
10 mg |
183.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18771518
|
Tocris Bioscience™
5319/50 |
50 mg |
629.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent, dual orexin receptor antagonist (IC50 values are 11 and 60 nM for OX2 and OX1 receptors respectively). Exhibits anxiolytic-like properties in vivo. Brain penetrant.Specifications
>99% | |
ACT 462206 | |
388.48 | |
ACT 462206 |
For Research Use Only