missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide/Bisacrylamide 29:1, powder

Premixed powder for preparing protein DNA gels

44.92€ - 207.03€

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 29:1
Physical Form Powder
Color White
MDL Number MFCD00080848
UN Number UN2074
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15443529
View Documents
Thermo Scientific Chemicals
J60824.17
40 g
44.92€
40g
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15453529
View Documents
Thermo Scientific Chemicals
J60824.22
100 g
82.21€
100g
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15463529
View Documents
Thermo Scientific Chemicals
J60824.28
200 g
207.03€
200g
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

Solubility
It is soluble in water.

Notes
Keep away from light. Store at room tempeature.
Specifications

Specifications

Acrylamide/Bisacrylamide 29:1
White
UN2074
Powder
MFCD00080848
It is soluble in water.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c

H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE, SOLID

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only