missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acid Fuchsin
Chemical Identifiers
CAS | 3244-88-0 |
---|---|
Molecular Formula | C20H17N3Na2O9S3 |
Molecular Weight (g/mol) | 587.544 |
MDL Number | MFCD00013286 |
InChI Key | VCEFFPMSOGDMJX-UHFFFAOYSA-N |
Synonym | Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S |
PubChem CID | 131852436 |
IUPAC Name | 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium |
SMILES | CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3244-88-0 | |
587.544 | |
VCEFFPMSOGDMJX-UHFFFAOYSA-N | |
131852436 | |
CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] |
C20H17N3Na2O9S3 | |
MFCD00013286 | |
Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S | |
3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium |
Specifications
3244-88-0 | |
C20H17N3Na2O9S3 | |
15, 99 | |
Solubility in water: 143g/L. Other solubilities: slightly soluble in alcohol | |
CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] | |
587.544 | |
585.52 | |
Green | |
Acid Fuchsin, Pure |
55% min. | |
MFCD00013286 | |
Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S | |
VCEFFPMSOGDMJX-UHFFFAOYSA-N | |
3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium | |
131852436 | |
Pure | |
Glittering Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acid Fuchsin
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only