missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone oxime, 98%

Product Code. 11449393
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11449393

Brand: Thermo Scientific Alfa Aesar A11804.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 613-91-2 | C8H9NO | 135.17 g/mol

Used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.

Solubility
Slightly soluble in water. Soluble in ethyl alcohol almost transparency.

Notes
Air and Moisture Sensitive. Store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-91-2
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00013931 MFCD00013931
InChI Key JHNRZXQVBKRYKN-VQHVLOKHSA-N
Synonym acetophenone oxime, ethanone, 1-phenyl-, oxime, acetophenone, oxime, methyl phenyl ketone oxime, methyl phenyl ketoxime, 1-phenylethanone oxime, z-acetophenone oxime, n-1-phenylethylidene hydroxylamine, e-n-1-phenylethylidene hydroxylamine, ne-n-1-phenylethylidene hydroxylamine
PubChem CID 5464950
IUPAC Name (NE)-N-(1-phenylethylidene)hydroxylamine
SMILES C\C(=N/O)C1=CC=CC=C1

Specifications

Melting Point 58°C to 60°C
Boiling Point 118°C to 120°C (20 mmHg)
Quantity 50 g
Beilstein 1562059
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in water. Soluble in ethyl alcohol almost transparency.
Formula Weight 135.16
Percent Purity 98%
Chemical Name or Material Acetophenone oxime

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.