missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, for GC Plus, Honeywell Burdick & Jackson™
Click to view available options
18 L
Specifications
Specifications
Melting Point | -95°C |
CAS | 67-64-1 |
Color | Colorless |
Density | 0.79 g/cm3 (20°C) |
Boiling Point | 55°C to 57°C (1013 hPa) |
Quantity | 18 L |
Flash Point | -17°C |
Molecular Formula | C3H6O |
MDL Number | MFCD00008765 |
Beilstein | 635680 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'