Learn More
Description
Acetic acid is a clear, colorless solution with a strong sour odor. It is the second simplest carboxylic acid (after formic acid) and consists of two small functional groups, an acetyl group and a hydroxyl group. Undiluted, it is referred to as “glacial” acetic acid.
Acetic acid is an important chemical reagent and industrial chemical for the production of chemical compounds. It is used in the production of photographic film, adhesives, paints, and synthetic fibers and fabrics. It is a primary component in the production of vinyl acetate monomer, as well as acetic anhydride and ester production. The volume of acetic acid used in vinegar is comparatively small.
Specifications
Specifications
pH | <2 |
CAS | 64-19-7, 7732-18-5 |
Color | Colorless |
Molecular Formula | C2H4O2 |
MDL Number | MFCD00036152 |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
SMILES | CC(O)=O |
Concentration | 12% by volume |
IUPAC Name | acetic acid |
Show More |
Safety and Handling


- Acetic acid 12 vol. % solution
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Acetic acid glacial
Your input is important to us. Please complete this form to provide feedback related to the content on this product.