Learn More
Fisherbrand™ AccuDuo Cartridge
Ensures optimum quality of the final product water. Fisher Scientific™ AccuDuo Cartridges provide dual column filtration for Accu20, Accu100, and Accu500 water systems.
272.00€ - 530.00€
Specifications
For Use With (Application) | Ultrapure water for critical laboratory applications |
---|
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
15459719
|
Fisherbrand™
|
H Pack | Accu500 |
361.00€
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
15449719
|
Fisherbrand™
|
P Pack | Accu20 |
272.00€
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
15489729
|
Fisherbrand™
|
U Pack | Accu20, Accu500 |
380.00€
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
15499729
|
Fisherbrand™
|
U Pack with a 0.2μm PES Capsule Filter | Accu20, Accu500 |
530.00€
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
15409739
|
Fisherbrand™
|
U Pack (low TOC) | Accu20, Accu500 |
468.00€
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
Description
- Includes two sets of dual-column purification cartridges
- Cartridges work sequentially to ensure thorough removal of ions and organics in water
Specifications
Ultrapure water for critical laboratory applications |