Learn More
Fisher Scientific™ Accu500 Water System
Product Code. 15459699 Shop All Fisherbrand Products
Click to view available options
Type:
UV Ultrapure Water System
UV Ultrapure Water System w/ Dispenser
Ultrapure Water Purification System
RO Flow Rate:
10 L/hr.
15 L/hr.
15 L/min.
20 L/hr.
30 L/hr.
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15459699
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
Exceeds ASTM, CLSI, CAP, and ISO Type I water standards. Fisher Scientific™ Accu500 Water System is a versatile purification system that can be used in a number of ways.
  • The fully automated control system contains pre-rinse, a flush feature preventing RO membrane fouling, RO membrane and cartridge life monitoring as well as many other programs for convenient operation
  • The pre-rinsing program starts automatically once a new RO membrane is installed so there is no need for user to set additional cleaning procedures
  • Two sets of dual-column purification cartridges work sequentially to ensure thorough removal of ions and organics in water
  • Automatic hourly system recirculation prevents bacterial growth
  • Resistivity is measured by a highly sensitive resistivity sensor
  • Electrical and water compartments are completely segregated, thus safer to operate
  • User can set water dispensing time for easy water collection
  • Both RO and ultrapure water can be dispensed from the water system
  • The system can be fitted with a 30L or 60L PE tank with a liquid level sensor and a tank vent filter with CO2 scavenger to protect purified water from atmospheric degradation
  • A 0.2μm filter or ultrafiltration filter (optional) removes final contaminants
  • Independent ultrapure water dispenser
  • The remote dispenser has a built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
  • Volumetric dispensing and calibration
  • The handle is 360 degree-rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility
Applications:
Ultrapure and RO water can be used in many areas. Some typical applications include:
Ultrapure water
  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.
RO water
  • Glassware cleaning
  • Washing machine for glassware
  • Waterbath water
  • Autoclaving
Specifications
Conductivity <20μS/cm
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Water Type Tap Water
Resistivity 18.2 Mohm.cm
Type UV Ultrapure Water System
Feedwater Pressure 1 to 6 bar
Model Accu500
Particles >0.2µM/mL <1/mL with 0.2μm final filter or terminal ultrafiltration cartridge
Purified Water Flowrate Field 1.5 L/min.
RO Flow Rate 20 L/hr.
Temperature Range (Metric) 5°C to 45°C
Length (Metric) 30 cm
Height (Metric) 48 cm
Depth (Metric) 51 cm