Learn More
Cole-Parmer™ Accessories for Cole-Parmer™ HI-200 Series Hybridization Incubators
Product Code. p-200004266
Click to view available options
Type:
Tube Rack Holder
Unit Size:
Each
Product Code. 11533859
Brand: Cole-Parmer™ 3945527
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Complement your hybridization incubation systems
  • Expand the capabilities of your Cole Parmer HI-200 Series Hybridization Incubators with our collection of optional accessories
  • This collection includes a selection of rocking platforms, hybridization tubes of various sizes, and tube holders with clips
Specifications
For Use With (Equipment) Hybridization incubator
Holds Up to 3 Large and 3 Small Tubes
Material Plastic
Type Tube Rack Holder
Includes Hybridization Tube Rack Holder
Length (Metric) 80 mm