Learn More
Thermo Scientific ACCESS PORT RIGHT DIA.50MMthe center of the right side wall D = 58 mm
Brand:  Thermo Scientific 51900999
ACCESS PORT RIGHT DIA.50MMthe center of the right side wall D = 58 mm
Product Code. 11824423
  • ONLINE EXCLUSIVE PRICE
    493.00€
    471.00€ / Each
    Save 22.00€
    4% Off
Only null left
Estimated Shipment: 04-02-2026